Rhenium(VII) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Rhenium(VII) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium(VII) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Moisture Sensitive; Sublimation temperature/start: 250°C (482°F)
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011339.04
also known as 011339-04
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
Add to cart
Rhenium(VII) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number011339.04
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-68-7
IUPAC Nametrioxo[(trioxorhenio)oxy]rhenium
Molecular FormulaO7Re2
InChI KeyNBGOSNNAAHDRLK-UHFFFAOYSA-N
SMILESO=[Re](=O)(=O)O[Re](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to black
Total Metal Impurities≤0.01% (100 ppm)
FormCrystalline
Rhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only