Niobium(V) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Niobium(V) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

For Use As An Intermediate and in Electronics
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 011365.A3
also known as 011365-A3
Price (USD)/ Each
1,432.65
Online exclusive
1592.00 
Save 159.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
1,432.65
Online exclusive
1592.00 
Save 159.35 (10%)
Add to cart
Niobium(V) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number011365.A3
Price (USD)/ Each
1,432.65
Online exclusive
1592.00 
Save 159.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1313-96-8
IUPAC Name[(dioxoniobio)oxy]dioxoniobium
Molecular FormulaNb2O5
InChI KeyZKATWMILCYLAPD-UHFFFAOYSA-N
SMILESO=[Nb](=O)O[Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
As an intermediate and in electronicsNiobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in the food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only