Cadmium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Cadmium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011367.FF
also known as 011367-FF
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
Add to cart
Cadmium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011367.FF
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium
Molecular FormulaCd
InChI KeyBDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 2.0mm (0.08in) thick.
Appearance (Color)White silvery
Assay from Supplier's CofA≥99.998% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only