Aluminum foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: Approx. 6.75g/50 x 50mm; Temper: as rolled
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
100 x 500 mm
Catalog number 011373.GH
also known as 011373-GH
Price (USD)/ Each
169.00
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
169.00
Add to cart
Aluminum foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011373.GH
Price (USD)/ Each
169.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormFoil
Comment0.9 - 1.1 mm thick
Total Metal Impurities0.002% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only