Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis), Thermo Scientific Chemicals
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis), Thermo Scientific Chemicals

Form: Approx. 2.7g/100 x 100mm; Temper: as rolled
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 100 x 400 mm
100 x 100 mm
100 x 400 mm
Catalog number 011376.IP
also known as 011376-IP
Price (USD)/ Each
897.00
-
Add to cart
Quantity:
5 x 100 x 400 mm
Request bulk or custom format
Price (USD)/ Each
897.00
Add to cart
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis), Thermo Scientific Chemicals
Catalog number011376.IP
Price (USD)/ Each
897.00
-
Add to cart
Specifications
Chemical Name or MaterialAluminum
ColorSilver
CAS7429-90-5
AppearanceSilver-colored foil
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only