Indium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Indium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 011382.FI
also known as 011382-FI
Price (USD)/ Each
395.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
395.00
Add to cart
Indium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011382.FI
Price (USD)/ Each
395.00
-
Add to cart
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.998% (metals basis)
FormFoil, 2.0mm (0.08in) thick.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only