Thermo Scientific Chemicals

Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011391.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011391.FI
50 x 50 mm, Each
Quantity
Catalog number: 011391.FI
also known as 011391-FI
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-57-5
IUPAC Name
gold
Molecular Formula
Au
InChI Key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES
[Au]
Comment
Gold
Form
Foil
Total Metal Impurities
≤ 0.0025%
Thickness
0.09-0.11mm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text