Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals
Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals

Form: approx. 1.2g/25x25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 011391.GH
also known as 011391-GH
Price (USD)/ Each
3,787.65
Online exclusive
4208.00 
Save 420.35 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
3,787.65
Online exclusive
4208.00 
Save 420.35 (10%)
Add to cart
Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals
Catalog number011391.GH
Price (USD)/ Each
3,787.65
Online exclusive
4208.00 
Save 420.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentGold
FormFoil
Total Metal Impurities≤ 0.0025%
Thickness0.09-0.11mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only