Thermo Scientific Chemicals

Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals

Catalog number: 011395.A1
1 kg, Each
Thermo Scientific Chemicals

Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals

Catalog number: 011395.A1
1 kg, Each
Quantity
Catalog number: 011395.A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-23-4
IUPAC Name
dioxozirconium
Molecular Formula
O2Zr
InChI Key
MCMNRKCIXSYSNV-UHFFFAOYSA-N
SMILES
O=[Zr]=O
Impurity content
HfO₂ 2% max
Total Metal Impurities excluding Hf
1% max.

Description

Zirconium(IV) oxide is used in the production of piezoelectric crystals and also used in the high frequency induction coils and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text