Thermo Scientific Chemicals

Calcium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011397.A3
2 kg, Each
Thermo Scientific Chemicals

Calcium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011397.A3
2 kg, Each
Quantity
Catalog number: 011397.A3
also known as 011397-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12049-50-2
IUPAC Name
titanium(4+) calcium trioxidandiide
Molecular Formula
CaO3Ti
InChI Key
WEUCVIBPSSMHJG-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Ca++].[Ti+4]
Total Metal Impurities
1% max.

Description

Calcium titanium oxide is used to prepare the ferrotitanium alloys and its structure is used to study the other compounds such as barium titanate.It is used to make the pure substances at low temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text