Thermo Scientific Chemicals

Calcium carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011403.22
100 g, Each
Thermo Scientific Chemicals

Calcium carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011403.22
100 g, Each
Quantity
Catalog number: 011403.22
also known as 011403-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-34-1
IUPAC Name
calcium carbonate
Molecular Formula
CCaO3
InChI Key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C([O-])=O
Particle Size
5 Micron average
Appearance (Color)
White
Total Metal Impurities
≤ 0.5%
Form
Powder

Description

Calcium carbonate is used in the purification of iron, refining of sugar from sugar beet and flue gas desulfurization applications in fossil fuel power stations. It is used in blackboard chalk, printing and writing paper, paints, plastics, PVC cables, polypropylene compounds, thermosetting resins, adhesives, sealants and decorating fillers. It is an abrasive and an ingredient in many glazes. It is also used in the pharmaceutical industry as an inert filler for tablets and other pharmaceuticals. It is used as a base in many organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text