Silver powder, crystalline, -10+20 mesh, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Silver powder, crystalline, -10+20 mesh, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver powder, crystalline, -10+20 mesh, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 011409.18
also known as 011409-18
Price (USD)/ Each
405.65
Online exclusive
450.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
405.65
Online exclusive
450.00 
Save 44.35 (10%)
Add to cart
Silver powder, crystalline, -10+20 mesh, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number011409.18
Price (USD)/ Each
405.65
Online exclusive
450.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver-grey
FormPowder
Total Metal Impurities≤0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only