Thermo Scientific Chemicals

Silver nitrate, ACS, 99.9+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011414.14
25 g, Each
Thermo Scientific Chemicals

Silver nitrate, ACS, 99.9+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011414.14
25 g, Each
Quantity
Catalog number: 011414.14
also known as 011414-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7761-88-8
IUPAC Name
silver(1+) nitrate
Molecular Formula
AgNO3
InChI Key
SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES
[Ag+].[O-][N+]([O-])=O
Assay (unspecified)
≥ 99.0% AgNO₃
Impurity content
Substances not precipitated by hydrochloric acid ≤ 0.01%
Solution Test
Clarity of solution P.T.
Form
Crystalline
Appearance (Color)
White to gray

Description

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate is used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text