Thermo Scientific Chemicals

Silver phosphate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011415.14
25 g, Each
Thermo Scientific Chemicals

Silver phosphate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011415.14
25 g, Each
Quantity
Catalog number: 011415.14
also known as 011415-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-09-0
IUPAC Name
trisilver(1+) phosphate
Molecular Formula
Ag3O4P
InChI Key
FJOLTQXXWSRAIX-UHFFFAOYSA-K
SMILES
[Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O
Appearance (Color)
Pale yellow to yellow
Total Metal Impurities
≤1.0%
Form
Powder

Description

Silver phosphate finds applications in photography, and in silver staining of biological materials. It is used as a photocatalyst for the visible light photochemical splitting of water, and for production of activated oxygen by the same method. It is employed in Crystal growth for scintillation, optics and dosimetry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text