Thermo Scientific Chemicals

Silver carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011420.22
100 g, Each
Thermo Scientific Chemicals

Silver carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011420.22
100 g, Each
Quantity
Catalog number: 011420.22
also known as 011420-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-16-7
IUPAC Name
carbonic acid silver
Molecular Formula
CH2AgO3
InChI Key
LKZMBDSASOBTPN-UHFFFAOYSA-N
SMILES
[Ag].OC(O)=O
Total Metal Impurities
0.5% max.

Description

Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text