Silver chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Silver chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 011421.22
also known as 011421-22
Price (USD)/ Each
546.65
Online exclusive
607.00 
Save 60.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
546.65
Online exclusive
607.00 
Save 60.35 (10%)
Add to cart
Silver chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011421.22
Price (USD)/ Each
546.65
Online exclusive
607.00 
Save 60.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-90-6
IUPAC Namesilver(1+) chloride
Molecular FormulaAgCl
InChI KeyHKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES[Cl-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Total Metal Impurities≤0.1% (1000 ppm)
Assay (unspecified)75.3 ? 0.4% Ag w/w
FormCrystalline solid
Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only