Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 x 200 mm
50 x 50 mm
100 x 100 mm
Catalog number 011440.GW
also known as 011440-GW
Price (USD)/ Each
256.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
302.00 
Save 45.35 (15%)
-
Add to cart
Quantity:
200 x 200 mm
Request bulk or custom format
Price (USD)/ Each
256.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
302.00 
Save 45.35 (15%)
Add to cart
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011440.GW
Price (USD)/ Each
256.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
302.00 
Save 45.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormFoil, 0.127mm (0.005in) thick, annealed.
Assay from Supplier's CofAAg + Cu: 99.94 - 100.0% (metals basis)
Assay from Supplier's CofAAl ≤ 20ppm
Assay from Supplier's CofABi ≤ 10ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only