Iron rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Iron rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Form: Approx.0.155g/mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 150 mm
50 mm
150 mm
Catalog number 011442.HT
also known as 011442-HT
Price (USD)/ Each
2,065.65
Online exclusive
2295.00 
Save 229.35 (10%)
-
Add to cart
Quantity:
5 x 150 mm
Request bulk or custom format
Price (USD)/ Each
2,065.65
Online exclusive
2295.00 
Save 229.35 (10%)
Add to cart
Iron rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number011442.HT
Price (USD)/ Each
2,065.65
Online exclusive
2295.00 
Save 229.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialIron
ColorGray
CAS7439-89-6
Health Hazard 1H500
AppearanceGray rod
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only