Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Form: approx. 32.94g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
500 cm
Catalog number 011465.BY
also known as 011465-BY
Price (USD)/ Each
274.65
Online exclusive
305.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (USD)/ Each
274.65
Online exclusive
305.00 
Save 30.35 (10%)
Add to cart
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number011465.BY
Price (USD)/ Each
274.65
Online exclusive
305.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Diameter1.8-2.2mm
CommentSilver
Total Metal Impurities≤ 0.001%
FormWire
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only