Silver rod, 7mm (0.275in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Silver rod, 7mm (0.275in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver rod, 7mm (0.275in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mm
100 mm
Catalog number 011473.DH
also known as 011473-DH
Price (USD)/ Each
117.00
-
Add to cart
Quantity:
25 mm
Request bulk or custom format
Price (USD)/ Each
117.00
Add to cart
Silver rod, 7mm (0.275in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number011473.DH
Price (USD)/ Each
117.00
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormRod, 7mm (0.275in) diameter.
Assay from Supplier's CofA≥99.999% (metals basis)
Assay from Supplier's CofA25mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only