Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.76g/cm
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 15 cm
15 cm
Catalog number 011478.QY
also known as 011478-QY
Price (USD)/ Each
449.00
-
Add to cart
Quantity:
5 x 15 cm
Request bulk or custom format
Price (USD)/ Each
449.00
Add to cart
Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Catalog number011478.QY
Price (USD)/ Each
449.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 6mm (0.2in) diameter
Assay from Supplier's CofA≥99.9965% (metals basis)
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only