Thermo Scientific Chemicals

Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 011484.36
500 g, Each
Thermo Scientific Chemicals

Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 011484.36
500 g, Each
Quantity
Catalog number: 011484.36
also known as 011484-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16872-11-0
IUPAC Name
hydrogen tetrafluoroboranuide
Molecular Formula
BF4H
InChI Key
ODGCEQLVLXJUCC-UHFFFAOYSA-O
SMILES
[H+].F[B-](F)(F)F
Appearance (Color)
Clear colorless to pale yellow
Assay from Suppliers CofA
≥48.0% to ≤51.0% (Titration)
Comment
May develop some turbidity or precipitate
Form
Solution

Description

Tetrafluoroboric acid is used in conjunction with various methods of cleaning and treat the surface of substrates. It is used as radiotracers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text