Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals
Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011487.09
also known as 011487-09
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
Add to cart
Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals
Catalog number011487.09
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-50-8
IUPAC Nameiron(3+) trifluoride
Molecular FormulaF3Fe
InChI KeySHXXPRJOPFJRHA-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min
Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only