Thermo Scientific Chemicals

Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals

Catalog number: 011487.18
50 g, Each
Thermo Scientific Chemicals

Iron(III) fluoride, anhydrous, 97% min, Thermo Scientific Chemicals

Catalog number: 011487.18
50 g, Each
Quantity
Catalog number: 011487.18
also known as 011487-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-50-8
IUPAC Name
iron(3+) trifluoride
Molecular Formula
F3Fe
InChI Key
SHXXPRJOPFJRHA-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Fe+3]
Assay (unspecified)
97% min

Description

Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text