Thermo Scientific Chemicals

Copper(II) fluoride, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog number: 011489.18
50 g, Each
Thermo Scientific Chemicals

Copper(II) fluoride, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog number: 011489.18
50 g, Each
Quantity
Catalog number: 011489.18
also known as 011489-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-19-7
IUPAC Name
copper(2+) difluoride
Molecular Formula
CuF2
InChI Key
GWFAVIIMQDUCRA-UHFFFAOYSA-L
SMILES
[F-].[F-].[Cu++]
Assay (unspecified)
99.5% min

Description

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent.Copper(II) fluoride is used in the preparation of fluorobenzene by reacting with benzene. It is also used in galvanic cells, high-temperature fluorinating agent and fluxing iron castings. Further, it is used as an efficient catalyst in Chan-Lam-Evans coupling reaction to synthesis the diaryl- ether compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text