Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals
Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals

Ammonium hexafluorophosphate, 99.5%, Alfa Aesar, Quantity: 100g, CAS: 16941-11-0, Melting Point: 198 deg.C, Molecular Formula: F6H4NP, Molecular Weight: 163.003
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011495.14
also known as 011495-14
Price (USD)/ Each
57.65
Online exclusive
64.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
64.20 
Save 6.55 (10%)
Add to cart
Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals
Catalog number011495.14
Price (USD)/ Each
57.65
Online exclusive
64.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
CAS16941-11-0
IUPAC Nameammonium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6H4NP
InChI KeyNIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES[NH4+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% min
Ammonium hexafluorophosphate is used as a preservative. It is used as a starting material in the preparation of other hexafluorophosphates. Further, it is used in synthesis of organometalic compounds, especially rhodicinium hexafluorophosphate, which has inert and non-coordinating counter ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only