Thermo Scientific Chemicals

Aluminum fluoride trihydrate, 97%, Thermo Scientific Chemicals

Catalog number: 011496.36
500 g, Each
Thermo Scientific Chemicals

Aluminum fluoride trihydrate, 97%, Thermo Scientific Chemicals

Catalog number: 011496.36
500 g, Each
Quantity
Catalog number: 011496.36
also known as 011496-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15098-87-0
IUPAC Name
aluminium(3+) trihydrate trifluoride
Molecular Formula
AlF3H6O3
InChI Key
BKLZIAYVINRQEJ-UHFFFAOYSA-K
SMILES
O.O.O.[F-].[F-].[F-].[Al+3]
Assay (unspecified)
96.1% min

Description

Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text