Thermo Scientific Chemicals

Aluminum fluoride, anhydrous, 99+%, Thermo Scientific Chemicals

Catalog number: 011497.18
50 g, Each
Thermo Scientific Chemicals

Aluminum fluoride, anhydrous, 99+%, Thermo Scientific Chemicals

Catalog number: 011497.18
50 g, Each
Quantity
Catalog number: 011497.18
also known as 011497-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-18-1
IUPAC Name
aluminium(3+) trifluoride
Molecular Formula
AlF3
InChI Key
KLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Al+3]
Assay (unspecified)
99.1% min

Description

In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactionsAluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text