Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals
Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals

Neutral; S.A. 150m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011502.A1
also known as 011502-A1
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
Add to cart
Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals
Catalog number011502.A1
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
pH6.5-7.5

Aluminum oxide is used as an abrasive, dosimeter, filler of choice for plastics, ingredient in sunscreen and cosmetics. Aluminum oxide flakes are also used in paint for providing reflective decorative effects. In laboratories it is widely used as a medium for chromatographic purifications. It is used in the production of aluminum (Hall–Héroult process), alumina nanofibers, cutoff tools, and sandpaper. It serves as a catalyst for several reactions of industrial importance, including converting alcohols into alkenes, decomposing hydrazine, drying of gases, and converting hydrogen sulfide into sulfur in refineries (Claus Process). It is also used in integrated circuits, compact fluorescent lamps (CFL), sodium vapor lamps, hip replacements, CD/DVD polishing, superconducting quantum interference devices (SQUIDS), insulation devices, and as a support for many popular catalysts (for example, in Ziegler-Natta polymerizations).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only