Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx.0.75g/25 x 25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011515.FF
also known as 011515-FF
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
Add to cart
Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011515.FF
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver gray
Assay from Supplier's CofA≥99.9% (metals basis)
FormFoil, 0.1mm (0.004in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only