Gold Thinfoil, 0.01mm (0.0004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Gold Thinfoil, 0.01mm (0.0004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold Thinfoil, 0.01mm (0.0004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011518.FF
also known as 011518-FF
Price (USD)/ Each
267.00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
267.00
Add to cart
Gold Thinfoil, 0.01mm (0.0004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011518.FF
Price (USD)/ Each
267.00
-
Add to cart
Specifications
Chemical Name or MaterialGold Thinfoil
Name Note0.01mm (0.0004 in.) thick, not certified pinhole free
CAS7440-57-5
Recommended StorageAmbient temperatures
EINECS Number231-165-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only