Iridium foil, 1.0mm (0.04in) thick, typically 99.7+% (metals basis), Thermo Scientific Chemicals
Iridium foil, 1.0mm (0.04in) thick, typically 99.7+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium foil, 1.0mm (0.04in) thick, typically 99.7+% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 x 25 mm
Catalog number 011525.FF
Price (USD)/ Each
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialIridium foil
Name Note1.0mm (0.04 in.) thick
CAS7439-88-5
Recommended StorageAmbient temperatures
EINECS Number231-095-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only