Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 011528.18
also known as 011528-18
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
Add to cart
Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Catalog number011528.18
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14283-07-9
IUPAC Namelithium(1+) tetrafluoroboranuide
Molecular FormulaBF4Li
InChI KeyUFXJWFBILHTTET-UHFFFAOYSA-N
SMILES[Li+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Lithium tetrafluoroborate is extensively utilized as an electrolyte in primary and secondary cells. It is used in rechargeable Li ion batteries. The electrolytes are non aqueous solutions of lithium tetrafluoroborate in the organic media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only