Thermo Scientific Chemicals

Mercury(II) fluoride, 95%, Thermo Scientific Chemicals

Catalog number: 011533.09
10 g, Each
Thermo Scientific Chemicals

Mercury(II) fluoride, 95%, Thermo Scientific Chemicals

Catalog number: 011533.09
10 g, Each
Quantity
Catalog number: 011533.09
also known as 011533-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-39-3
IUPAC Name
mercury(2+) difluoride
Molecular Formula
F2Hg
InChI Key
FMSYTQMJOCCCQS-UHFFFAOYSA-L
SMILES
[F-].[F-].[Hg++]
Assay (unspecified)
94.1% min

Description

Mercury(II) fluoride is a selective fluorination agent. Organic compounds, such as triphenylacetic acid, triphenyl ethylene, and triethyl phosphite can be selectively fluorinated in dimethylsulfoxide/HgF 2 solutions under UV-visible illumination. CF2I2 was synthesized by fluorination of tetraiodomethane with mercury(II) fluoride and purified.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text