Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals
Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 011537.A1
also known as 011537-A1
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
Add to cart
Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals
Catalog number011537.A1
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7488-55-3
IUPAC Nameλ²-tin(2+) sulfate
Molecular FormulaO4SSn
InChI KeyOBBXFSIWZVFYJR-UHFFFAOYSA-L
SMILES[Sn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95.5% min
It is used in tin plating baths and it is a precursor for tin metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only