Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals
Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 011538.03
also known as 011538-03
Price (USD)/ Each
25.65
Online exclusive
27.90 
Save 2.25 (8%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
27.90 
Save 2.25 (8%)
Add to cart
Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals
Catalog number011538.03
Price (USD)/ Each
25.65
Online exclusive
27.90 
Save 2.25 (8%)
-
Add to cart
Chemical Identifiers
CAS26042-64-8
Molecular FormulaAgF6Sb
InChI KeyVWZIPIHZVCRLOX-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[F].[Ag+].[Sb]
Molecular Weight (g/mol)343.62
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.5% (based on Silver Elemental Analysis)
Elemental AnalysisFluorine, F: Reported
Elemental AnalysisSilver, Ag: Reported
Silver hexafluoroantimonate is used as a catalyst for the preparation of a number of compounds. It acts as an acidic catalyst in epoxide opening reactions and sulfenylation reactions , controlled initiator in polymerization reactions, and as an activator of catalyst in certain organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only