Thermo Scientific Chemicals

Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals

Catalog number: 011538.14
25 g, Each
Thermo Scientific Chemicals

Silver hexafluoroantimonate, 99%, Thermo Scientific Chemicals

Catalog number: 011538.14
25 g, Each
Quantity
Catalog number: 011538.14
also known as 011538-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26042-64-8
Molecular Formula
AgF6Sb
InChI Key
VWZIPIHZVCRLOX-UHFFFAOYSA-N
SMILES
[F].[F].[F].[F].[F].[F].[Ag+].[Sb]
Molecular Weight (g/mol)
343.62
Appearance (Color)
White to pale cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.5% (based on Silver Elemental Analysis)
Elemental Analysis
Fluorine, F: Reported
Elemental Analysis
Silver, Ag: Reported

Description

Silver hexafluoroantimonate is used as a catalyst for the preparation of a number of compounds. It acts as an acidic catalyst in epoxide opening reactions and sulfenylation reactions , controlled initiator in polymerization reactions, and as an activator of catalyst in certain organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text