Thermo Scientific Chemicals

Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals

Catalog number: 011541.18
50 g, Each
Thermo Scientific Chemicals

Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals

Catalog number: 011541.18
50 g, Each
Quantity
Catalog number: 011541.18
also known as 011541-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-49-5
IUPAC Name
zinc(2+) difluoride
Molecular Formula
F2Zn
InChI Key
BHHYHSUAOQUXJK-UHFFFAOYSA-L
SMILES
[F-].[F-].[Zn++]
Assay (unspecified)
99% min

Description

It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text