Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals
Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 011541.30
also known as 011541-30
Price (USD)/ Each
288.65
Online exclusive
320.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
288.65
Online exclusive
320.00 
Save 31.35 (10%)
Add to cart
Zinc fluoride, anhydrous, 99% min, Thermo Scientific Chemicals
Catalog number011541.30
Price (USD)/ Each
288.65
Online exclusive
320.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-49-5
IUPAC Namezinc(2+) difluoride
Molecular FormulaF2Zn
InChI KeyBHHYHSUAOQUXJK-UHFFFAOYSA-L
SMILES[F-].[F-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only