Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals
Zirconium(IV) fluoride, low hafnium, 99%, Hf &lt;0.1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011542.22
also known as 011542-22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7783-64-4
IUPAC Namezirconium(4+) tetrafluoride
Molecular FormulaF4Zr
InChI KeyOMQSJNWFFJOIMO-UHFFFAOYSA-J
SMILES[F-].[F-].[F-].[F-].[Zr+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Impurity contentHf <0.1%
Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only