Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals
Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 011543.14
also known as 011543-14
Price (USD)/ Each
233.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
233.00
Add to cart
Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals
Catalog number011543.14
Price (USD)/ Each
233.00
-
Add to cart
Chemical Identifiers
CAS10049-16-8
IUPAC Namevanadium pentafluoride
Molecular FormulaF5V
InChI KeyLYTSFXQJUMZNFC-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only