Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals
Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 011544.22
also known as 011544-22
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
Add to cart
Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals
Catalog number011544.22
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
-
Add to cart
Chemical Identifiers
CAS7783-47-3
IUPAC Nameλ²-tin(2+) difluoride
Molecular FormulaF2Sn
InChI KeyANOBYBYXJXCGBS-UHFFFAOYSA-L
SMILES[F-].[F-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder or crystals
Assay from Suppliers CofA≥97.5% (Titration)
Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only