Thermo Scientific Chemicals

Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals

Catalog number: 011544.36
500 g, Each
Thermo Scientific Chemicals

Tin(II) fluoride, 97.5%, Thermo Scientific Chemicals

Catalog number: 011544.36
500 g, Each
Quantity
Catalog number: 011544.36
also known as 011544-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-47-3
IUPAC Name
λ²-tin(2+) difluoride
Molecular Formula
F2Sn
InChI Key
ANOBYBYXJXCGBS-UHFFFAOYSA-L
SMILES
[F-].[F-].[Sn++]
Appearance (Color)
White to pale cream
Form
Powder or crystalline powder or crystals
Assay from Suppliers CofA
≥97.5% (Titration)

Description

Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text