Thermo Scientific Chemicals

Aluminum nitride, N 32.0% min, Thermo Scientific Chemicals

Catalog number: 011546.18
50 g, Each
Thermo Scientific Chemicals

Aluminum nitride, N 32.0% min, Thermo Scientific Chemicals

Catalog number: 011546.18
50 g, Each
Quantity
Catalog number: 011546.18
also known as 011546-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24304-00-5
IUPAC Name
alumanylidyneamine
Molecular Formula
AlN
InChI Key
PIGFYZPCRLYGLF-UHFFFAOYSA-N
SMILES
N#[Al]
Assay (unspecified)
32.0% N min

Description

Aluminum nitride possesses very high thermal conductivity and effectively used in the ceramic industry. It finds application in deep ultraviolet optoelectronics, steel production, dielectric layers in optical storage media, chip carriers and as a crucible to grow crystals of gallium arsenide. It is also used as a radio frequency filter, which finds application in mobile phones. Further, it is used as a circuit carrier in semiconductors. In addition to this, it is used as a heat-sink in light-emitting diode lighting technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text