Thermo Scientific Chemicals

Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011548.14
25 g, Each
Thermo Scientific Chemicals

Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011548.14
25 g, Each
Quantity
Catalog number: 011548.14
also known as 011548-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-12-7
IUPAC Name
niobium(5+) pentachloride
Molecular Formula
Cl5Nb
InChI Key
YHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
Appearance (Color)
Yellow
Form
Powder
Total Metal Impurities
≤0.1% (1000 ppm)
Particle Size
-100 Mesh

Description

It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text