Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 011548.36
also known as 011548-36
Price (USD)/ Each
811.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
811.00
Add to cart
Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011548.36
Price (USD)/ Each
811.00
-
Add to cart
Chemical Identifiers
CAS10026-12-7
IUPAC Nameniobium(5+) pentachloride
Molecular FormulaCl5Nb
InChI KeyYHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.1% (1000 ppm)
Particle Size-100 Mesh
Appearance (Color)Yellow
It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only