Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Catalog number 011551.18
also known as 011551-18
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number011551.18
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS12039-88-2
IUPAC Nametungsten disilicide
Molecular FormulaSi2W
InChI KeyWQJQOUPTWCFRMM-UHFFFAOYSA-N
SMILES[Si]#[W]#[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Tungsten silicide is used in microelectronics as a contact material. It is also used as a shunt over polysilicon lines to increase their conductivity and increase signal speed. Further, it acts as a barrier layer between silicon and other metals. In addition to this, it is used in microelectro mechanical systems and for oxidation-resistant coatings. It is also employed as a replacement for earlier tungsten films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only