Thermo Scientific Chemicals

Sodium carbonate, anhydrous, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 011552.18
50 g, Each
Thermo Scientific Chemicals

Sodium carbonate, anhydrous, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 011552.18
50 g, Each
Quantity
Catalog number: 011552.18
also known as 011552-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
497-19-8
IUPAC Name
disodium carbonate
Molecular Formula
CNa2O3
InChI Key
CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C([O-])=O
Assay (unspecified)
99.5% Na₂CO₃ min
Comment
Loss on heating at 285?C 1.0%
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Ca 0.03%
Impurity content
Cl 0.001%

Description

Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text