Thermo Scientific Chemicals

Potassium hydrogen fluoride, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011557.A3
2 kg, Each
Thermo Scientific Chemicals

Potassium hydrogen fluoride, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011557.A3
2 kg, Each
Quantity
Catalog number: 011557.A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-29-9
IUPAC Name
potassium hydrogen difluoride
Molecular Formula
F2HK
InChI Key
VBKNTGMWIPUCRF-UHFFFAOYSA-M
SMILES
[H+].[F-].[F-].[K+]
Total Metal Impurities
0.9% max.

Description

Potassium hydrogen fluoride is widely used as an etchant in glass and cleaning products. It finds an important application in the synthesis of special optical glass viz. crown and crown flint glass and as a catalyst for polymerization. It is involved in the manufacturing of wood preservatives, soldering agents and brazing. It is also used in the manufacturing of organic and inorganic fluorine compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Electrolyte, flux, catalyst in alkylation of benzene with olefins, and in the synthesis of pure KF
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text