Thermo Scientific Chemicals

Zinc acetate dihydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 011559.30
250 g, Each
Thermo Scientific Chemicals

Zinc acetate dihydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 011559.30
250 g, Each
Quantity
Catalog number: 011559.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5970-45-6
IUPAC Name
zinc(2+) diacetate dihydrate
Molecular Formula
C4H10O6Zn
InChI Key
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES
O.O.[Zn++].CC([O-])=O.CC([O-])=O
Impurity content
Fe 5ppm
Impurity content
K 0.01%
Impurity content
Na 0.05%
Impurity content
Pb 0.002%
Impurity content
SO₄ 0.005%

Description

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text